Learn More
Meridian Bioscience™ Alethia™ Incubator Reader
GSA_VA
Catalog No. 23029014 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
1
Catalog No. Quantity
23-029-014 1
1 options
This item is not returnable. View return policy
Catalog No. 23-029-014 Supplier Meridian Bioscience™ Supplier No. 610189
Add to Cart
Add to Cart
This item is not returnable. View return policy
Meridian Bioscience™ Alethia™ Incubator Reader designed for performing Alethia™ Assays.
  • An automated isothermal amplification and detection system for performing Alethia™ assays
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Isothermal Amplification and Detection System
For Use With (Application) Alethia™ Assays
Quantity 1