Learn More
Vector Laboratories Aleuria Aurantia Lectin (AAL), Biotinylated, 1mg
SDP
Supplier:  Vector Laboratories B13951
Encompass_Preferred
1 mg. Immunohistochemistry / Immunocytochemistry, Immunofluorescence, Blotting Applications, Elispot, ELISAs, Glycobiology.Recommended Storage2-8 °C; Store frozen for long term storage.Inhibiting and/or Eluting Sugar100-400 mM L-fucose.Solution10 mM HEPES, pH 7.5, 0.15 M NaCl, 0.08% sodium azide.Concentration2 mg active conjugate/ml
Catalog No. NC1877241
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.