Learn More
ACROBiosystems Alexa Fluor 488-Labeled Human
Supplier:  ACROBiosystems HLDHA2H4200UG
Encompass_Preferred
Alexa Fluor 488-Labeled Human HLA-A*11:01&B2M&KRASG12D (VVVGADGVGK) Complex Protein (HLD-HA2H4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*11:01) & Ile 21 - Met 119 (B2M) & VVVGADGVGK peptide (Accession # Q5S3G3-1 (HLA-A*11:01) & P61769 (B2M) & VVVGADGVGK).
Catalog No. 50-253-4735
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.