Learn More
Biolegend Alexa Fluor 647 anti-c-Myc
Supplier:  Biolegend 626810
Encompass_Preferred
Alexa Fluor 647 anti-c-Myc [9E10]; Isotype: Mouse IgG1; Reactivity: Endogenous Human c-Myc; fusion protein with c-Myc tag (EQKLISEEDL, around 1.2kD) in all species; Apps: ICC; Size: 100 ug
Catalog No. 50-208-6837
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.