Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alexa Fluor™ 647 IgG1, κ N297A Human Isotype Control, BD Pharmingen™

Catalog No. BDB569794 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569794 25 μg
BDB569961 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569794 Supplier BD Biosciences Supplier No. 569794
Add to Cart
Add to Cart

Human Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry, Control
Classification Monoclonal
Clone X40.huIgG1N297A.rMAb (also known as X40.huIgG1 N297A.rMAb)
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IgG1, kappa Isotype Control (Anti-KLH)
Host Species Human
Immunogen Keyhole limpet hemocyanin (KLH)
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Content And Storage The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.