Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alexa Fluor™ 647 Mouse IgG1 κ Isotype Control, BD Pharmingen™

Catalog No. BDB570232 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570232 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB570232 Supplier BD Biosciences Supplier No. 570232
Add to Cart
Add to Cart

Isotype Control

It is validated for Intracellular staining (flow cytometry)|Isotype control
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry, Control
Clone MOPC-21 (also known as MOPC21; MOPC 21)
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IgG1, kappa; Ighg1, Igh-4, γg1; Igk-C, IGKC, Ig κ
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.