Learn More
Cayman Chemical AlexamorelIntrIfluoroACTAt 5mg
SDP
Supplier:  Cayman Chemical 365865 MG
Encompass_Preferred
A natriuretic peptide increases ex vivo A23187-induced vasorelaxation in porcine hearts isolated from a model of acute coronary occlusion at 2 Ug/kg bolus dose followed by 0 01 Ug/kg per minute infusion
Catalog No. 50-291-3906
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.