Learn More
Thermo Scientific Chemicals Polyethylene Glove Bag, 68cm (27in) x 43cm (17in) x 30cm (12in), 56cm (22in) entrance
For use with handling air/moisture sensitive high purity chemicals or materials. Ideal for laboratory use and field sampling.
$119.60 - $714.65
Specifications
| Material | Polyethylene |
|---|---|
| Color | Clear |
| For Use With (Application) | Field Sampling, Laboratory Use |
| Catalog Number | Mfr. No. | Dimensions (L x W x H) | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W x H) | Quantity | Price | Quantity & Availability | ||||
|
AA91717LK
|
Thermo Scientific Chemicals
091717LK |
17 x 17 x 11 in. | 1 Ea. |
Each for $119.60
Order in Bulk
Request bulk or custom formats |
|
||||
|
AA92717LK
|
Thermo Scientific Chemicals
092717LK |
27 x 17 x 12 in. | 1 Ea. |
$166.80
$133.65 / Each Save $33.15 20% Off
Order in Bulk
Request bulk or custom formats |
|
||||
|
AA92717LL
|
Thermo Scientific Chemicals
092717LL |
27 x 17 x 12 in. | 4 Pk. |
Each for $591.50
Order in Bulk
Request bulk or custom formats |
|
||||
|
AA92727LK
|
Thermo Scientific Chemicals
092727LK |
27 x 27 x 15 in. | 1 Ea. |
$198.70
$171.65 / Each Save $27.05 14% Off
Order in Bulk
Request bulk or custom formats |
|
||||
|
AA92727LL
|
Thermo Scientific Chemicals
092727LL |
27 x 27 x 15 in. | 4 Pk. |
$705.00
$636.65 / Each Save $68.35 10% Off
Order in Bulk
Request bulk or custom formats |
|
||||
|
AA93727LK
|
Thermo Scientific Chemicals
093727LK |
37 x 27 x 18 in. | 1 Ea. |
Each for $173.50
Order in Bulk
Request bulk or custom formats |
|
||||
|
AA93727LL
|
Thermo Scientific Chemicals
093727LL |
37 x 27 x 18 in. | 4 Pk. |
$580.50
$518.65 / Each Save $61.85 11% Off
Order in Bulk
Request bulk or custom formats |
|
||||
|
AA93737LK
|
Thermo Scientific Chemicals
093737LK |
37 x 37 x 25 in. | 1 Ea. |
$276.00
$240.65 / Each Save $35.35 13% Off
Order in Bulk
Request bulk or custom formats |
|
||||
|
AA93737LL
|
Thermo Scientific Chemicals
093737LL |
37 x 37 x 25 in. | 4 Pk. |
$889.00
$714.65 / Each Save $174.35 20% Off
Order in Bulk
Request bulk or custom formats |
|
||||
Description
Polyethylene Glove Bag is used for handling the air and moisture sensitive chemicals in laboratories. It is also used for field sampling and minimizes work exposure. Further, it preserves the integrity of the sample during storage. In addition to this, it serves as a portable laboratory for conducting tests and crime scene investigations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A portable alternative to standard glove boxes
- Includes closure, tubing coupler, and cotton gloves
- Ideal for handling air/moisture sensitive, high purity chemicals or materials
- For use in the laboratory and/or field sampling
- Available in a variety of sizes and quantities
Specifications
| Polyethylene | |
| Clear | |
| Field Sampling, Laboratory Use |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.