Learn More
Alfa Aesar™ Polyethylene Glove Bags
For handling air and moisture sensitive, high purity chemicals and materials
Manufacturer:  Alfa Aesar 93737LK
 View more versions of this product
Catalog No. AA93737LK
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
94cm x 94cm x 64cm; 88cm entrance
1/bag
For handling air, moisture sensitive, high purity chemicals and materials