Learn More
Apexbio Technology LLC Alfacalcidol 41294-56-8 500mg
SDP
Supplier:  Apexbio Technology LLC B2153500
Encompass_Preferred
Alfacalcidol is a vitamin D analog functioning as an inhibitor of bone resorption through modulation of calcium homeostasis By binding specifically to vitamin D receptors (VDR) located in calcium-regulating tissues such as intestine bone kidney and parathyroid glands Alfacalcidol contributes to mineral metabolism and bone turnover regulation Preclinical studies using ovariectomized animal models demonstrate that Alfacalcidol at pharmacological doses not inducing hypercalcemia suppresses osteoclast-mediated bone resorption while preserving calcium balance in plasma and urine Due to these properties Alfacalcidol is frequently utilized in biomedical research to investigate mechanisms involved in osteoporosis progression and potential therapeutic interventions addressing calcium and bone metabolism disorders
Catalog No. 50-000-62298
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.