Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA1031 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1031 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA1031 Supplier Invitrogen™ Supplier No. PA1031
  • $596.37
    $461.65 / Each of 1

    Save $134.72

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

ALG1 Polyclonal Antibody for Western Blot

The enzyme encoded by this gene catalyzes the first mannosylation step in the biosynthesis of lipid-linked oligosaccharides. This gene is mutated in congenital disorder of glycosylation type Ik.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALG1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene alg-1
Gene Accession No. 0
Gene Alias Alg1; alg-1; ALG1 chitobiosyldiphosphodolichol beta-mannosyltransferase; ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase; Argonaute; Argonaute-like; asparagine-linked glycosylation 1 (beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1 homolog (S. cerevisiae, beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1 homolog (yeast, beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1, beta-1,4-mannosyltransferase homolog; asparagine-linked glycosylation protein 1 homolog; beta-1,4 mannosyltransferase; beta-1,4-mannosyltransferase; CDG1K; CELE_F48F7.1; chitobiosyldiphosphodolichol beta-mannosyltransferase; GDP-Man:GlcNAc2-PP-dolichol mannosyltransferase; GDP-mannose-dolichol diphosphochitobiose mannosyltransferase; HMAT1; hMat-1; HMT1; HMT-1; Mannosyltransferase-1; Mat-1; MT-1; Protein argonaute; protein-coding; PSEC0061; UNQ861/PRO1870
Gene Symbols alg-1
Host Species Rabbit
Immunogen Peptide sequence DLEEIPNSPPTQPQTFSDVPQR.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 181504
Target Species C. elegans
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.