Learn More
ALG1L2 Polyclonal Antibody

Description
ALG1L2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | ALG1L2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | ALG1L2 |
| Gene Accession No. | C9J202 |
| Gene Alias | ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase like 2; ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase-like 2; ALG1L2; asparagine-linked glycosylation 1-like 2; putative glycosyltransferase ALG1L2 |
| Gene Symbols | ALG1L2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.