Learn More
Research Products International Corp ali-Q
SDP
Supplier:  Research Products International Corp 163260VS
Encompass_Preferred
Unique ali-Q 2 Pipet Controller offers sensitive touch-control aspirate and dispense buttons with repeat pipeting using serological pipets. Features include easy to use aliquot volume dial; aliquot button; exceptional repeatability; innovative system of sensors electronically controlled valves, advanced computer functions for added accuracy and precision; 2 Multifunction LED indicators; and multiple alert tones.
Catalog No. 50-238-8427
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.