Learn More
VistaLab™ Technologies ali-Q 2 Maintenace Parts and Accessories
Shop All VistaLab Technologies Inc. Products

Description
- For use with the ali-Q and/or ali-Q 2 Aliquoting Pipet Controller
Specifications
Specifications
| Autoclavable | Autoclavable |
| Disposable | Single-use |
| For Use With | ali-Q 2 Aliquoting Pipet Controller |
| Material | PTFE |
| Pore Size | 0.22 μm |
| Sterility | Non-sterile |
| Type | Nozzle Filter |
| Filter Type | 0.22 μm PTFE Membrane Filter |
| For Use With (Equipment) | ali-Q or ali-Q 2 pipet controller |
| Quantity | 5/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.