Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AquaPhoenix Scientific Aligator Clip Leads, BLK
SCI_ED

Catalog No. S02900 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S02900 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02900 Supplier AquaPhoenix Scientific Supplier No. AC5001BLK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reliable 12 in. black alligator clip leads designed for quick, secure circuit testing in educational and lab environments.

  • 12 in. (30.5 cm) insulated wire with alligator clips on both ends
  • Durable black insulation for clear circuit identification
  • Ideal for temporary electrical connections and testing
  • Perfect for use in science labs, classroom experiments, and electronics kits
  • Flexible, reusable, and easy to handle

Specifications

Product Type Alligator Clip Leads
For Use With (Equipment) Circuits
Length (English) 12 in.
Color Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.