Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alignflow™ Flow Cytometry Alignment Beads for Red Lasers, 6.0 μm

Catalog No. A16504
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16504 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16504 Supplier Invitrogen™ Supplier No. A16504
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Alignflow™ Flow Cytometry Alignment Beads for Red Lasers, 6.0 μm

Molecular Probes™ AlignFlow™ flow cytometry alignment beads are reliable references for aligning, focusing, and calibrating flow cytometers.The 6.0μm Alignflow™ Flow Cytometry Alignment Beads for Red Lasers are fluorescent-dye infused microspheres that are excitable at 630–640 nm with emission in the range of 645–680 nm.

View information for all flow cytometry calibration and size reference beads.

AlignFlow™ beads provide:

  • Uniformity—very low FSC CV and intensity CV
  • Consistency in production, creating ideal reference standards
  • Confidence through reliable, optimal instrument performance
  • Compatibility—use with any instrument

    The AlignFlow™ beads are highly uniform with respect to size and fluorescence intensity, resulting in low FSC CV and intensity CV. The beads approximate the size, emission wavelength, and intensity of many biological samples and permit the calibration of the flow cytometer’s laser source, optics, and stream flow without wasting valuable and sensitive experimental material.

    Bead Labeling
    As with all of our flow cytometry standard microspheres, the AlignFlow™ beads are stained internally rather than on the surface. The dyes are therefore insulated from environmental interactions that could cause variable fluorescence output, resulting in excellent signal stability. These fluorescent polystyrene microspheres are supplied as suspensions packaged in dropper vials for convenient dispensation and are available in a choice of three colors (UV, blue, red) and two sizes (2.5μm, 6.0μm) to match your laser.

    Notes:
    The AlignFlow™ flow cytometry reference beads are provided in updated packaging that includes a Quick Reference Label that puts key information for product use at your fingertips.

    These beads replace previous AlignFlow™ flow cytometry reference bead products. This particular product replaces AlignFlow™ Plus Flow Cytometry Alignment Beads, 6.0μm, for 633 excitation (A7313).
  • TRUSTED_SUSTAINABILITY

    Specifications

    Diameter (Metric) 6 μm
    Content And Storage Contains 2 vials of Alignflow™ beads (1.5 mL per vial). Store at 2–8°C.
    Format Suspension
    For Use With (Equipment) Flow Cytometer
    Quantity 3 mL
    Detection Method Fluorescence
    Excitation Wavelength Range 633 or 635
    Stain Configuration Core
    Shipping Condition Room Temperature
    Product Line AlignFlow
    Product Type Alignment Beads
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.