Learn More
Cayman Chemical 14TrIamcInolon acetonIde 500UG
Supplier: Cayman Chemical 9000087500 UG
A Nampt activator increases NMN levels and prevents NAD feedback inhibition in a cell-free assay using recombinant human Nampt at 2 UM increases liver levels of NAD but does not affect NAD levels in the heart quadriceps or gastrocnemius in mice at 20 mg/kg
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.