Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alizarin-3-Methyliminodiacetic Acid, 85%, Solstice

Catalog No. 60044572 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-572 5 g Glass Bottle
60-044-571 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-572 Supplier Solstice Supplier No. 055905G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for the determination of fluorine, ∼85%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3952-78-1
Molecular Formula C19H15NO8
Molecular Weight (g/mol) 385.328
MDL Number MFCD00001202
InChI Key PWIGYBONXWGOQE-UHFFFAOYSA-N
Synonym alizarin complexone, alizarin fluorine blue, alizarin complexon, alizarine complexon, alizarine complexone, alizarinkomplexon, alizarine fluorine blue, 3-aminomethylalizarin-n,n-diacetic acid, az-c, alizarin-3-methyliminodiacetic acid
PubChem CID 65132
ChEBI CHEBI:53088
IUPAC Name 2-[carboxymethyl-[(3,4-dihydroxy-9,10-dioxoanthracen-2-yl)methyl]amino]acetic acid
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)CN(CC(=O)O)CC(=O)O

Specifications

Melting Point 185°C
Color Yellow
Packaging Glass Bottle
Quantity 5 g
UN Number NONH for all modes of transport
Beilstein 2190028
Formula Weight 385.32g/mol
Percent Purity 85%
Chemical Name or Material Alizarin-3-Methyliminodiacetic Acid
EINECSNumber 223-544-2
RTECSNumber AH0585000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.