accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHOOOOHOHO

CAS RN 3952-78-1

IUPAC Name: 2-[(carboxymethyl)[(3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracen-2-yl)methyl]amino]acetic acid
Synonyms: alizarin complexone alizarine fluorine blue alizarin fluorine blue alizarinkomplexon alizarin complexon alizarine complexone alizarine complexon Az-C
Molecular Weight (g/mol): 385.33
Molecular Formula: C19H15NO8
InChi Key: PWIGYBONXWGOQE-UHFFFAOYSA-N
SMILES: OC(=O)CN(CC(O)=O)CC1=CC2=C(C(O)=C1O)C(=O)C1=CC=CC=C1C2=O
Quantity 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (2)
Color 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)