Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Alizarin, 97%, pure

Catalog No. AC153691000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153691000 100 g Glass bottle
AC153690250 25 g Glass bottle
AC153695000 500 g Glass bottle
AC153690025 2.5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC153691000 Supplier Thermo Scientific Chemicals Supplier No. 153691000
Add to Cart

CAS: 72-48-0 | C14H8O4 | 240.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-48-0
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.214
MDL Number MFCD00001201
InChI Key RGCKGOZRHPZPFP-UHFFFAOYSA-N
Synonym 1, 2-Dihydroxyanthraquinone, Mordant Red 11
PubChem CID 6293
ChEBI CHEBI:16866
IUPAC Name 1,2-dihydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3)O)O

Specifications

Melting Point 287.0°C to 289.0°C
Boiling Point 430.0°C
CAS Min % 96.0
CAS Max % 100.0
Dye Content 96.0% min.
Assay Percent Range 97%
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in aromatic solvents, ether and hot, methanol, moderately soluble in ethanol
Infrared Spectrum Authentic
Formula Weight 240.21
Loss on Drying 2% max.
Percent Purity 97%
Grade Pure
Packaging Glass bottle
Quantity 100 g
Physical Form Fine Powder
Chemical Name or Material Alizarin, 97%
Show More Show Less
Hazard Category Warning
EINECSNumber 200-782-5
RTECSNumber CB6580000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.