Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alizarin Complexone 95.0+%, TCI America™
SDP

Catalog No. A04401G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A04401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A04401G Supplier TCI America Supplier No. A04401G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3952-78-1
Molecular Formula C19H15NO8
Molecular Weight (g/mol) 385.328
MDL Number MFCD00001202
InChI Key PWIGYBONXWGOQE-UHFFFAOYSA-N
Synonym alizarin complexone, alizarin fluorine blue, alizarin complexon, alizarine complexon, alizarine complexone, alizarinkomplexon, alizarine fluorine blue, 3-aminomethylalizarin-n,n-diacetic acid, az-c, alizarin-3-methyliminodiacetic acid
PubChem CID 65132
ChEBI CHEBI:53088
IUPAC Name 2-[carboxymethyl-[(3,4-dihydroxy-9,10-dioxoanthracen-2-yl)methyl]amino]acetic acid
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)CN(CC(=O)O)CC(=O)O

Specifications

Melting Point 180°C
Quantity 1 g
Formula Weight 385.33
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Alizarin Complexone
RTECSNumber AH0585000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.