Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alizarin complexone, MP Biomedicals™

Catalog No. ICN203429 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN203429 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN203429 Supplier MP Biomedicals, Inc Supplier No. 0520342901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alizarin complexone is suitable for the colorimetric detection of fluorine. Alizarin complexone is also known that it has antiretroviral activity for the medication purpose of the treatment of infection by retroviruses

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3952-78-1
Molecular Formula C19H15NO8
Molecular Weight (g/mol) 385.328
MDL Number MFCD00001202
InChI Key PWIGYBONXWGOQE-UHFFFAOYSA-N
Synonym Alizarin-3-methylamine-N, N-diacetic acid
PubChem CID 65132
ChEBI CHEBI:53088
IUPAC Name 2-[carboxymethyl-[(3,4-dihydroxy-9,10-dioxoanthracen-2-yl)methyl]amino]acetic acid
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)CN(CC(=O)O)CC(=O)O

Specifications

Color Brown
Quantity 1 g
Physical Form Fine Powder
Formula Weight 385.34
Grade Not Available
Chemical Name or Material Alizarin complexone
Recommended Storage Store at Room Temperature (15 to 30°C).
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.