Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ ALK (Anaplastic Lymphoma Kinase)/p80 Ab-1 Mouse Monoclonal Antibody, 7mL

Catalog No. p-4536138 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1104S0 100 μL Concentrated
MS1104S 1 mL Concentrated
MS1104S1 500 μL Concentrated
MS1104R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1104S0 Supplier Epredia™ Supplier No. MS1104S0
Only null left
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ ALK (Anaplastic Lymphoma Kinase)/p80 Ab-1, Mouse Monoclonal Antibody.

ALK/p80 is a hybrid of the anaplastic lymphoma kinase (ALK) gene and the nucleophosmin (NPM) gene resulting from the t(2;5)(p23;q35) translocation found in a third of large cell lymphomas.

Host Species: Mouse

Clone: 5A4

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Epitope: aa 419-520 (tyrosine kinase domain)

Immunogen: Recombinant protein corresponding to a region which spans the tyrosine kinase catalytic domain and part of the C-terminus of the NPM-ALK transcript (419-520aa)

Molecular Weight: 80kDa

Positive Control: Anaplastic lymphoma

Cellular Localization: Cytoplasmic and/or nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not recommended)

Specifications

Antigen ALK (Anaplastic Lymphoma Kinase)/p80 Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5A4
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to a region which spans the tyrosine kinase catalytic domain and part of the C-terminus of the NPM-ALK transcript (419-520aa)
Quantity 100 μL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.