Learn More
Description
Use alkaline batteries for high-use applications like:
- MP3 Players
- Disc Players
- Cameras
- Handheld TVs
- Handheld Games
- Stereos
- Remote Controlled Devices
Specifications
Specifications
| Product Type | Alkaline Battery |
| Battery Type | C |
| For Use With (Application) | For peak performance in high-tech electronic devices and high-use devices |
| For Use With (Equipment) | For MP3 players, disc players, cameras, handheld TVs, handheld games, stereos, remote controlled devices |
| Quantity | 2/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
