Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkaline Phosphatase, 2500 U, G-Biosciences
SCI_ED

Catalog No. S001107
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001107 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001107 Supplier G-Biosciences Supplier No. 7862267
Add to Cart
Add to Cart

TAB5 Alkaline Phosphatase is a heat labile alkaline phosphatase. It catalyzes the dephosphorylation of phosphomonoesters at both 5' and 3' ends of DNA and RNA molecules in a nonspecific manner. Additionally, it eliminates phosphate groups from the 5' and 3' positions of DNA, RNA, and dNTPs.

  • This enzyme finds utility in various molecular biology applications, including the dephosphorylation of DNA and RNA ends to prepare them for cloning or probe end-labeling.

Specifications

Packaging Vial
Enzyme Alkaline Phosphatase
Sensitive to Heat Inactivation Yes
Product Type TAB5 Alkaline Phosphatase
Research Category Signal Transduction, Drug Development, Genome Editing
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.