Learn More
Alkaline Phosphatase/ALPP Antibody (8B6), PE, R&D Systems™

Description
Specifications
Specifications
| Antigen | Alkaline Phosphatase/ALPP |
| Applications | ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Radioimmunodiffusion, Western Blot |
| Classification | Monoclonal |
| Clone | 8B6 |
| Conjugate | PE |
| Dilution | Flow Cytometry |
| Gene Alias | Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme |
| Gene Symbols | ALPP |
| Host Species | Mouse |
| Immunogen | Hep-2 cells with boosted surface expression of Alkaline Phosphatase, Placental. [UniProt# P05187]. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.