Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Alkaline phosphatase (Chicken)

Catalog No. 89933183 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-183 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-183 Supplier Abnova™ Supplier No. P5247
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Name Alkaline phosphatase (Chicken)
Preparation Method Native protein purified from Chicken Intestine
Quantity 250 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name ALKALINE
Assays Alkaline Phosphatase Assay; NF/USP dexamethasone phosphate assay.
Biological Activity ≥0.9 units per mg dry weight (25°C pH 8.8). One Unit hydrolyzes 1 umole of o-carboxyphenol phosphate per minute at 25°C, pH 8.8.
Species Chicken
Protein Tag None
Expression System Native protein purified from Chicken Intestine
Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.