Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Alkaline Phosphatase (E. coli)

Catalog No. 89933185 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-185 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-185 Supplier Abnova™ Supplier No. P5294
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Formulation In 2.6M ammonium sulfate, pH 8.0
Name Alkaline Phosphatase (E. coli)
Preparation Method Native protein chromatographically purified from E. coli
Quantity 5 mg
Storage Requirements Store at 4°C. This product is stable for at least 3 months.
Regulatory Status RUO
Common Name ALKALINE
Assays Alkaline Phosphatase Assay
Biological Activity ≥20 units per mg protein (25°C, pH 8.0). One Unit hydrolyzes 1 umole of p-nitrophenol phosphate per minute at 25°C, pH 8.0.
Species E. coli
Protein Tag None
Expression System Native protein chromatographically purified from E. coli
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.