Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkaline phosphatase from E. coli, MP Biomedicals™

Catalog No. MP219981794 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219981794 5 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219981794 Supplier MP Biomedicals, Inc Supplier No. 0219981794
Add to Cart
Add to Cart
Request a Quote

Alkaline phosphatase from E. coli ≥30 u/mg protein , Activity- ∽1000 u/mL, Suspension

  • It is used for conjugation to antibodies and other proteins for ELISA, Western blotting, and histochemical detection.
  • It may be used for protein labeling when high sensitivity is required.
  • Alkaline phosphatase is a hydrolase enzyme responsible for removing phosphate groups from many types of molecules, including nucleotides, proteins, and alkaloids.
  • It is most effective in an alkaline environment. It is sometimes used synonymously as basic phosphatase.
TRUSTED_SUSTAINABILITY

Specifications

pH 10.4
For Use With (Application) Cell Biology and Analysis, Molecular biology
Content And Storage Store at 4°C, do not freeze.
Source E.coli
Quantity 5 kU
Product Type Alkaline Phosphatase
Synonym Orthophosphoric Monoester Phosphohydrolase
Form Suspension
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.