Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Alkaline Phosphatase Enhancer, Signal Enhancer, 250 mL

Catalog No. 502710097
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-0097 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-0097 Supplier Abcam Supplier No. AB671250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Signal enhancer for alkaline phosphatase-based immunostaining systems.

  • Signal enhancer for use with alkaline phosphatase based immunostaining systems
  • Storage: Store at Ambient
  • Shipping: Ambient (can ship with ice)

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Alkaline Phosphatase
Shipping Condition Ambient
Form Liquid
Reactivity Alkaline Phosphatase
Detection Method Chromogenic
Product Type Luminescence Enhancer
Content And Storage Store at ambient temperature.
Format Bottle
Quantity 250 mL
For Use With (Application) Immunostaining
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.