Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Alkaline Protease (APA) for ReliaPrep™ Large Volume HT gDNA Isolation System

Catalog No. PRA1721 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
130 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA1721 130 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA1721 Supplier Promega Supplier No. A1721
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alkaline Protease (APA) is a component of the ReliaPrep Large Volume HT gDNA Isolation System (Cat.# A2751).

  • Reagent Used in DNA Isolation Protocols
  • Component of the ReliaPrep™ Large Volume HT gDNA Isolation System (Cat.# A2751) available as a standalone item

Specifications

Content And Storage 15°C to 30°C
Quantity 130 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.