Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkalinity Standard, 500 ppm, Reagents

Catalog No. 01335999
Change view
Click to view available options
Quantity:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-999 1 L
01-336-000 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-999 Supplier Reagents Holdings Llc Supplier No. CA0177001A
Add to Cart
Add to Cart

Alkalinity Standard, 500 ppm, Reagent, use for Water and wastewater analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Alkalinity Standard, 500 ppm
Packaging Poly Bottle
CAS 7732-18-5, 497-19-8
Specific Gravity 1.0003
Physical Form Liquid
Color Colorless
pH 10
Boiling Point 100°C
Solubility Completely miscible
Type Conductivity Standard
Quantity 1 L
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.