Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALKBH8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549590
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549590 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549590 Supplier Invitrogen™ Supplier No. PA549590
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALKBH8 Polyclonal Antibody for Western Blot, IHC (P), Flow

Catalyzes the methylation of 5-carboxymethyl uridine to 5-methylcarboxymethyl uridine at the wobble position of the anticodon loop in tRNA via its methyltransferase domain (PubMed:20123966, PubMed:20308323). Catalyzes the last step in the formation of 5-methylcarboxymethyl uridine at the wobble position of the anticodon loop in target tRNA (PubMed:20123966, PubMed:20308323). Has a preference for tRNA(Arg) and tRNA(Glu), and does not bind tRNA(Lys)(PubMed:20308323). Binds tRNA and catalyzes the iron and alpha-ketoglutarate dependent hydroxylation of 5-methylcarboxymethyl uridine at the wobble position of the anticodon loop in tRNA via its dioxygenase domain, giving rise to 5-(S)-methoxycarbonylhydroxymethyluridine; has a preference for tRNA(Gly) (PubMed:21285950). Required for normal survival after DNA damage (PubMed:20308323). May inhibit apoptosis and promote cell survival and angiogenesis (PubMed:19293182).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALKBH8
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ALKBH8
Gene Accession No. Q96BT7
Gene Alias 4930562C03Rik; 8030431D03Rik; 9430088N01Rik; ABH8; alkB homolog 8, tRNA methyltransferase; AlkB homologue 8; alkB, alkylation repair homolog 8; alkB, alkylation repair homolog 8 (E. coli); Alkbh8; Alkylated DNA repair protein alkB homolog 8; LOW QUALITY PROTEIN: alkylated DNA repair protein alkB homolog 8; probable alpha-ketoglutarate-dependent dioxygenase ABH8; RGD1304687; S-adenosyl-L-methionine-dependent tRNA methyltransferase ABH8; TRM9; TRMT9; tRNA (carboxymethyluridine(34)-5-O)-methyltransferase ABH8; tRNA methyltransferase 9 homolog
Gene Symbols ALKBH8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 494-523 amino acids from the C-terminal region of human ALKBH8.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91801
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.