Learn More
Sigma Aldrich Fine Chemicals Biosciences Alkyltrimethylammonium bro1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M76351KG
Encompass
Alkyltrimethylammonium bromide is a surfactant and can be used to study the forces and stability of foam films produced from soluble cationic surfactants.
Catalog No. 50-290-4222