Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam All-cis-7,10,13,16,19-Methyl Docosapentaenoate, >99% Purity, 25 mg
Description
- Molecular Weight: 344.5 Da
- Purity: >99%
- Description: Long chain polyunsaturated ω-3 fatty acid ester
- Potential Applications: Tumorigenesis-related studies, fatty acid studies, and internal standard
Specifications
Specifications
| Substrate Properties | Lipid-based Substrate |
| Target Molecule | Methyl docosapentaenoate |
| Shipping Condition | Blue Ice |
| Buffer | 100% all-cis-7,10,13,16,19-methyl docosapentaenoate |
| Content And Storage | Store at -20°C. |
| Formulation | 100% all-cis-7,10,13,16,19-Methyl docosapentae |
| Quantity | 25 mg |
| For Use With (Application) | Tumorigenesis-related studies, fatty acid studies, internal standard |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?