Learn More
Wells Lamont™ All Day™ Sleeve
GSA_VAGREENER_CHOICE
Catalog No. 17000484 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Style:
Sleeve with Thumb Hole
Standard Sleeve
Size:
14 in.
18 in.
24 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style Size
17-000-484 Standard Sleeve -
17-000-482 Standard Sleeve 14 in.
17-000-483 Standard Sleeve 18 in.
17-000-485 Sleeve with Thumb Hole -
17-000-486 Sleeve with Thumb Hole 24 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-000-484 Supplier Wells Lamont™ Supplier No. AD24
Add to Cart
Add to Cart
The All Day™ Sleeve offers ANSI A2 cut resistance and is made out of recycled water bottles. Features an inner cotton liner for comfort, elastic on both ends, and is latex free. Made in the USA.
  • Protect arms and the environment with cut resistant sleeves made of recycled material that would normally go to the landfill
  • High strength All Day™ fiber provides superior snag, tear, and abrasion resistance
  • Free of harsh dyes, chemicals, and latex
  • Lightweight construction provides all day comfort and fit
  • Elastic on both ends hold sleeve in place and cotton inner layer for added comfort
  • Three lengths are available depending on the length needed for protection
  • Available with or without a thumb hole
  • Not intended for use around sparks and flames
  • Made in the USA
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Cut-resistant Sleeve
Material Recycled Water Bottles
Sterility Non-sterile
Style Standard Sleeve
Disposable Single-use
Latex Free Yes