Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bausch & Lomb™ All-in-One Metal Lens-Cleaning Station
Designed for harsh or high-traffic work environments
Supplier: Bausch & Lomb™ 8569GM
Includes: Includes 1520 absorbent low-lint tissues to clean and buff eyewear and 16 oz. spray pump lens cleaning formula (silicone or non-silicone) for anti-fog protection in most conditions
Description
- Heavy-gauge blue metal cabinet with a low-maintenance enamel finish and a keylock for product retention
- Mounts easily with included fasteners
- Full drop-front access allows easy refilling
- Tissue disposal compartment keeps work area free of litter
- Measures 17.5L × 18.25W × 3.5 in.H (9 × 44 × 46cm)
Specifications
Anti-Fog Lens Cleaner w/out silicone,16 oz. | |
8569GMP metal station | |
16 oz. |
Cleaning solution | |
Non-silicone anti-fog formula | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction