Learn More
Chemglass Life Sciences Clamp Holder, All-Position
Supplier: Chemglass Life Sciences CG925701
Clamp Holder, All-Position. Double swivel design allows clamp holder to adjust at any angle in any plane. Holders are 90° to each other, joined by a right angle connector, allowing 360° rotation. Outside adjustment screws permit close proximity between items being held. Maximum rod grip size is 3/4” (19mm).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.