Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

all-trans-N-(4-Hydroxyphenyl)retinamide 98.0+%, TCI America™
SDP

Catalog No. H1464100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1464100MG 100 mg
H146410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1464100MG Supplier TCI America Supplier No. H1464100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65646-68-6
Molecular Formula C26H33NO2
Molecular Weight (g/mol) 391.555
MDL Number MFCD00792674
InChI Key AKJHMTWEGVYYSE-FXILSDISSA-N
Synonym fenretinide, n-4-hydroxyphenyl retinamide, 4-hpr, 4-hydroxyphenylretinamide, 4-hydroxyphenyl retinamide, retinoic acid p-hydroxyanilide, all-trans-4'-hydroxyretinanilide, fenretinida, fenretinidum, fenretinidum latin
PubChem CID 5288209
ChEBI CHEBI:42588
IUPAC Name (2E,4E,6E,8E)-N-(4-hydroxyphenyl)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenamide
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)NC2=CC=C(C=C2)O)C)C

Specifications

Melting Point 172°C
Quantity 100 mg
Formula Weight 391.56
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material all-trans-N-(4-Hydroxyphenyl)retinamide
RTECSNumber VH6420000
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.