Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ All Weather Permanent Adhesive Vinyl Valve Tag Labels for M6/M7 Printers

Catalog No. 17011523 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
1.5 x 2.875 in.
1.9 x 3.875 in.
Height (Metric):
38.1 mm
48.26 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Height (Metric)
17-011-523 1.9 x 3.875 in. 48.26 mm
17-011-519 1.5 x 2.875 in. 38.1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-011-523 Supplier Brady™ Supplier No. M62400595WT
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use durable, vinyl labels for indoor and outdoor identification. These glossy, pre-sized labels are a great way to mark valve tags.

  • Abrasion, chemical and oil-resistant
  • B-595 labels can be applied at cold temperatures
  • High gloss vinyl material has a permanent acrylic adhesive and is thermal transfer printable
  • For use with the M610 Label Printer and BradyPrinter M611 Mobile Label Printer
  • B-595 material has an 8 to 10 year average outdoor durability in temperatures from -40 to 180°F (-40 to 82°C); can be applied as low as 0°F (-18°C)

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance ANSI Z53.1-1967, ANSI Z535-2006, OSHA 1910.145, RoHS Compliant
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Gloss
Format Die-cut
Material Vinyl
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) General Identification, Pipe and Valve Marking, Electrical Labeling, Safety Identification, Hazard Communication Identification
For Use With (Equipment) M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
Latex Free No
Quantity 100 Labels
Shape Rectangle
Temperature Range (English) -40°F to +180°F
Temperature Range (Metric) -40°C to +82°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (L x W) 1.9 x 3.875 in.
Length (English) 1.9 in.
Width (English) 3.875 in.
Width (Metric) 98.43 mm
Height (Metric) 48.26 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.