Learn More
Allegro™ Ambient Air Driven Pump A‐4000AD, 4-Worker Connection
Catalog No. 19087454
Encompass_Preferred
Catalog No. 19-087454 Supplier Allegro™ Supplier No. 9850S
Portable air source for up to four full or half-face respirator users, or three hood users. Ideal for use when electric power is unavailable or undesirable.
  • For four respirator users or three hood users
  • Rigid metal base and housing
  • Brass and galvanized steel plumbing
  • Choose from hansen or schrader fittings
  • Adjustable brass pressure relief valve
  • Pressure gauge 0-15 psi (0-1.03 bar)
  • Oil-less, rotary vane, direct driven pump
  • Inlet filter 50 Micron felt material with metal grid inlet filter housing
  • Exhaust filter 0.3 Micron HEPA
Specifications
Material Metal base and housing, steel plumbing
Model 9850S
Weight (English) 83 lb.
Product Type Low Pressure Air Pump