Learn More
Allegro™ Ambient Air Pump A-1500TE, 3-Worker Connection
Catalog No. 19087460
Encompass_Preferred
Catalog No. 19-087460 Supplier Allegro™ Supplier No. 9832S
Ambient air pumps for up to three full or half-face respirator users, or two hood users. Fully assembled with all filters, fittings, and couplers with no need for additional monitoring and alarm systems.
  • Portable SAR air compressor, 3-worker connection
  • Hansen or Schrader Coupling
  • Rigid metal base and housing with four rubber feet
  • Brass and galvanized steel plumbing
  • Adjustable brass pressure relief valve
  • Motor 1.5 HP, 115V/230V motor
  • Pressure gauge 0-1 5 psi (0-1.03 bar)
Specifications
Applications Chemical Handling and Clean-up
For Use With (Equipment) Up to 3 respirators or 2 hoods
Material Metal base and housing, brass and steel plumbing
Model 9832S
Weight (English) 97 lb.
Product Type Low Pressure Air Pump