Learn More
Allegro™ Ambient Air Pump, Model A-300
Catalog No. 19087458
Encompass_Preferred
Catalog No. 19-087-458 Supplier Allegro™ Supplier No. 9806S
Oil-less ambient air systems for use in environments where an outside air source is required. Does not require carbon monoxide monitoring equipment. Choose from hansen or schrader fittings.
  • Rigid metal base and housing
  • Adjustable brass pressure relief valve
  • Oil-less, rotary vane pump
  • Motor 1/4 HP, 115V/230 VAC, Single Phase
  • Quick-connect hansen or schrader fitting
  • Inlet Filter 40.0 Micron bronze/steel
  • Exhaust Filter 0.30 Micron HEPA inline
Specifications
Applications Painting, Cleaning, Sanding, Chemical Handling
For Use With (Equipment) 1 full or half-face respirator
Material Metal base housing, brass plumbing
Model 9806S
Weight (English) 33 lb.
Product Type Low Pressure Air Pump