Learn More
Allegro™ Compressed Air Respirator Accessory, Personal Air Cooler with Belt and Hansen Fitting
Catalog No. 19999102
Encompass_Preferred
Change view
Click to view available options
Applications:
Hot Environments
Catalog No. Applications
19-999-102 Hot Environments
1 options
Catalog No. 19-999-102 Supplier Allegro™ Supplier No. 9991
Filtered compressed air source accessory changes the temperature of the delivered air to keep workers comfortable.
  • Aluminum
  • Hansen fitting 1/4 in. (63 cm) plug (series 1000) stainless steel
  • Nylon belt
Specifications
Certifications/Compliance NIOSH
Applications Hot Environments
For Use With (Equipment) Filtered Compressed Air Respirator
Material Aluminum Body and Leather Sheath
Includes Nylon waist belt and 0.25 in. Stainless steel Hansen quick-connect fitting
Model 9991
Pressure 55 to 85 psi
Product Type Air Cooler