Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allegro™ SAR Hood

Catalog No. 19169025
Change view
Click to view available options
Replacement hood; Color: white
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-169-025 Replacement hood; Color: white
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-169-025 Supplier Allegro™ Supplier No. 991139

Maintenance-free, disposable and convenient

  • One piece with breathing tube and head suspension integrated with hood assembly
  • Made of flashspun high-density polyethylene fibers
  • Splash-resistant and flexible

Specifications

Product Type Hood
Color White
Disposable Single-use
Material Poly coated Flashspun HDPE Fiber
For Use With (Equipment) Supplied air respirator
Includes Breathing tube, head suspension integrated with hood assembly
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.