Learn More
Allegro™ Respirator Cleaning Kit
Description
Includes
Supplied with 90 half-ounce (15g) packets of water-soluble cleaner, an air bulb, brush, 100 sterile storage bags with pressure-sensitive seals that can be marked with cleaning information, and a two-sided 17 qt. (16L) bucket.
Available with choice of a 32 oz. (0.9L) container of concentrated liquid disinfectant soap (EPA registered) or 90 half-ounce (15g) packets of cleaner.
Specifications
Specifications
| Product Type | Cleaning Kit |
| Model | 4001 |
| Includes | Supplied with ninety 0.5 oz. (15 g) packets of water soluble cleaner, an air bulb, brush, 100 sterile storage bags with pressure sensitive seals that can be marked with cleaning information and a two sided 17 qt. (16 L) bucket |
| For Use With (Equipment) | Full face and half mask respirator |
| Certifications/Compliance | OSHA |