Learn More
Allegro™ Respirator Cleaning Kit
Catalog No. 19088526
Change view
Click to view available options
Product Type:
Cleaning Kit
Cleaning Kit with Liquid Disinfectant Soap
Replacement Soap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-088-526 Cleaning Kit with Liquid Disinfectant Soap
19-088-600 Replacement Soap
19-087463 Cleaning Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-088-526 Supplier Allegro™ Supplier No. 4002
Add to Cart
Add to Cart
Includes
Supplied with 90 half-ounce (15g) packets of water-soluble cleaner, an air bulb, brush, 100 sterile storage bags with pressure-sensitive seals that can be marked with cleaning information, and a two-sided 17 qt. (16L) bucket.
All the necessary components to clean and store full facepiece and half-mask respirators in easy-to-order kits.
Available with choice of a 32 oz. (0.9L) container of concentrated liquid disinfectant soap (EPA registered) or 90 half-ounce (15g) packets of cleaner.
Specifications
Product Type Cleaning Kit with Liquid Disinfectant Soap
Model 4002
Includes Supplied with ninety 0.5 oz. (15 g) packets of water soluble cleaner, an air bulb, brush, 100 sterile storage bags with pressure sensitive seals that can be marked with cleaning information and a two sided 17 qt. (16 L) bucket
Certifications/Compliance OSHA