Learn More
TARGETMOL CHEMICALS INC Z-Semaxinib 5MG
Supplier: TARGETMOL CHEMICALS INC T24965MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. (Z)-Semaxinib (SU5416) is a potent and selective VEGFR(Flk-1/KDR) inhibitor (IC50 1.23 uM), 20-fold more selective for VEGFR over PDGFRbeta, no inhibition for FGFR, InsR, and EGFR. (Z)-Semaxinib is a quinolone derivative with potential antineoplastic activity. Purity 99.94%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.