Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Alligator Clip Leads
SDPSCI_ED

Catalog No. p-200009360 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (English):
12 in.
24 in.
36 in.
Catalog No. Length (English)
S15721 12 in.
S15881 36 in.
S15771 24 in.
3 options

Catalog No. S15721

Supplier: United Scientific™ WAG012PK/6

Only null left
Add to Cart
Add to Cart

Useful in a wide variety of electricity demonstrations.

  • Flexible, 18 gauge (SWG) stranded wire cord with an alligator clip on each end.
  • Alligator clip has a protective vinyl sheath.
  • Available in various lengths.
  • Packs of 6 (3 red and 3 black).

Specifications

Product Type Alligator Clip Leads
For Use With (Equipment) Electricity Experiment Supplies
Length (English) 12 in.
For Use With (Application) Electrical Experiments

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.