Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allitol 98.0+%, TCI America™
SDP

Catalog No. A1982100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1982100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1982100MG Supplier TCI America Supplier No. A1982100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity Allitol monosaccharide for use as a laboratory reagent.

Chemical Identifiers

CAS 488-44-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD11519461
InChI Key FBPFZTCFMRRESA-FBXFSONDSA-N
PubChem CID 120700
IUPAC Name (2R,3R,4S,5S)-hexane-1,2,3,4,5,6-hexol
SMILES OC[C@H](O)[C@H](O)[C@H](O)[C@H](O)CO

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 100 mg
Formula Weight 182.17
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Allitol
RTECSNumber BA1840000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.